1-(2-Chloro-5-fluoro-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
72341-500MG
72341-1G
72341-5G
72341-25G
77333-716EA
202.91
USD
77333-716
77357-888
77369-726
77390-664
1-(2-Chloro-5-fluoro-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Chloro-5-fluoro-3-methylphenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₅ClF₄O MW: 240.582 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34764317 CAS Number: 2807441-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...