5-Chloro-3-fluoro-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72304-500MG 72304-1G 72304-5G 72304-25G
77333-712EA 104.03 USD
77333-712 77343-062 77369-722 77381-788
5-Chloro-3-fluoro-2-isopropoxybenzoic acid ≥97%
5-Chloro-3-fluoro-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
MW: 232.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764285
CAS Number: 2469839-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR