2-Bromo-3-chloro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

72292-1G 72292-5G 72292-25G
77341-368EA 82.4 USD
77341-368 77369-716 77381-784
2-Bromo-3-chloro-4-(methylthio)pyridine ≥97%
2-Bromo-3-chloro-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNS
MW: 238.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764274
CAS Number: 2901099-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR