2-Fluoro-5-formyl-3-isopropoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
72280-500MG
72280-1G
72280-5G
72280-25G
77333-704EA
156.56
USD
77333-704
77345-626
77369-712
77390-656
2-Fluoro-5-formyl-3-isopropoxybenzoic acid ≥97%
2-Fluoro-5-formyl-3-isopropoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₁FO₄ Storage Temperature: Ambient |
MDL Number:
MFCD34764264 CAS Number: 2901102-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...