4-Fluoro-3-iodo-5-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72278-500MG 72278-1G 72278-5G 72278-25G
77333-702EA 110.21 USD
77333-702 77343-278 77369-710 77381-782
4-Fluoro-3-iodo-5-isopropoxybenzaldehyde ≥97%
4-Fluoro-3-iodo-5-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀FIO₂
MW: 308.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764262
CAS Number: 2901102-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR