4-Fluoro-2-iodo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72276-500MG 72276-1G 72276-5G 72276-25G
77333-700EA 201.88 USD
77333-700 77357-878 77369-708 77390-654
4-Fluoro-2-iodo-5-methoxybenzaldehyde ≥97%
4-Fluoro-2-iodo-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764260
CAS Number: 2387275-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR