5-Fluoro-2-formyl-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72273-500MG 72273-1G 72273-5G
77335-978EA 371.83 USD
77335-978 77357-876 77381-780
5-Fluoro-2-formyl-4-methoxybenzoic acid ≥97%
5-Fluoro-2-formyl-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764257
CAS Number: 2383640-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR