Methyl(3-methyl-5-propoxyphenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72268-500MG 72268-1G 72268-5G 72268-25G
77333-696EA 136.99 USD
77333-696 77344-632 77369-704 77390-650
Methyl(3-methyl-5-propoxyphenyl)sulfane ≥97%
Methyl(3-methyl-5-propoxyphenyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD34764253
CAS Number: 2807440-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR