1-Iodo-3-methyl-5-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

72267-500MG 72267-1G 72267-5G 72267-25G
77333-694EA 120.51 USD
77333-694 77343-812 77369-702 77381-778
1-Iodo-3-methyl-5-propoxybenzene ≥97%
1-Iodo-3-methyl-5-propoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃IO
MW: 276.1140 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764252
CAS Number: 2807467-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR