3-Methyl-5-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72265-500MG 72265-1G 72265-5G 72265-25G
77333-692EA 184.37 USD
77333-692 77347-334 77369-700 77390-648
3-Methyl-5-propoxybenzaldehyde ≥97%
3-Methyl-5-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.2276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764251
CAS Number: 2901106-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR