1-(4,5-Difluoro-2-methoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

72101-500MG 72101-1G 72101-5G
77336-964EA 400.67 USD
77336-964 77357-868 77381-770
1-(4,5-Difluoro-2-methoxyphenyl)-2,2-difluoroethanone ≥97%
1-(4,5-Difluoro-2-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103759
CAS Number: 1823320-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR