2,2-Difluoro-1-(4-fluoronaphthalen-1-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

72018-500MG 72018-1G 72018-5G 72018-25G
77333-688EA 182.31 USD
77333-688 77347-024 77369-692 77390-644
2,2-Difluoro-1-(4-fluoronaphthalen-1-yl)ethanone ≥97%
2,2-Difluoro-1-(4-fluoronaphthalen-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD28103916
CAS Number: 1690837-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR