1-(4-Chloro-2,5-dimethylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

72009-500MG 72009-1G 72009-5G 72009-25G
77333-686EA 178.19 USD
77333-686 77346-694 77369-690 77390-642
1-(4-Chloro-2,5-dimethylphenyl)-2,2-difluoroethanone ≥97%
1-(4-Chloro-2,5-dimethylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₂O
MW: 218.628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764226
CAS Number: 1694461-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR