1-(4-Bromo-2,5-dimethylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

72002-500MG 72002-1G 72002-5G 72002-25G
77333-684EA 182.31 USD
77333-684 77347-022 77369-688 77390-640
1-(4-Bromo-2,5-dimethylphenyl)-2,2-difluoroethanone ≥97%
1-(4-Bromo-2,5-dimethylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₂O
MW: 263.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29945630
CAS Number: 1698290-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR