2,2-Difluoro-1-(4-fluoro-2-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71978-500MG 71978-1G 71978-5G
77336-960EA 400.67 USD
77336-960 77357-860 77381-762
2,2-Difluoro-1-(4-fluoro-2-methoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(4-fluoro-2-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103884
CAS Number: 1824050-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR