1-(4-Chloro-2-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71943-500MG 71943-1G 71943-5G
77339-600EA 602.55 USD
77339-600 77357-856 77381-758
1-(4-Chloro-2-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone ≥97%
1-(4-Chloro-2-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30717162
CAS Number: 2169357-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR