1-(4-Bromo-2-methoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71919-500MG 71919-1G 71919-5G
77339-152EA 554.14 USD
77339-152 77357-852 77381-754
1-(4-Bromo-2-methoxyphenyl)-2,2-difluoroethanone ≥97%
1-(4-Bromo-2-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
CAS Number: 2228988-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR