1-(2-(Benzyloxy)phenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71901-500MG 71901-1G 71901-5G 71901-25G
77333-682EA 194.67 USD
77333-682 77357-850 77369-686 77390-638
1-(2-(Benzyloxy)phenyl)-2,2-difluoroethanone ≥97%
1-(2-(Benzyloxy)phenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₂
MW: 262.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764213
CAS Number: 2352757-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR