1-(3,4-Dichlorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71871-500MG 71871-1G 71871-5G 71871-25G
77333-680EA 210.12 USD
77333-680 77357-848 77369-684 77390-636
1-(3,4-Dichlorophenyl)-2,2-difluoroethanone ≥95%
1-(3,4-Dichlorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄Cl₂F₂O
MW: 225.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920638
CAS Number: 1271477-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR