2,2-Difluoro-1-(2-isopropoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71864-500MG 71864-1G 71864-5G
77336-956EA 400.67 USD
77336-956 77357-846 77381-752
2,2-Difluoro-1-(2-isopropoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-isopropoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045172
CAS Number: 1564904-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR