1-(4-Chloro-3-ethylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71840-500MG 71840-1G 71840-5G
77336-950EA 400.67 USD
77336-950 77357-840 77381-746
1-(4-Chloro-3-ethylphenyl)-2,2-difluoroethanone ≥97%
1-(4-Chloro-3-ethylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD28104181
CAS Number: 1822673-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR