1-(5-Chloro-2-hydroxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71791-500MG 71791-1G 71791-5G
77339-256EA 570.62 USD
77339-256 77357-836 77381-742
1-(5-Chloro-2-hydroxyphenyl)-2,2-difluoroethanone ≥95%
1-(5-Chloro-2-hydroxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O₂
MW: 206.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764205
CAS Number: 2228638-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR