2,2-Difluoro-1-(2-methoxy-6-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71773-500MG 71773-1G 71773-5G
77339-150EA 554.14 USD
77339-150 77357-832 77381-738
2,2-Difluoro-1-(2-methoxy-6-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-methoxy-6-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764196
CAS Number: 2229217-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR