2,2-Difluoro-1-(3-methoxy-4-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

71766-500MG 71766-1G 71766-5G 71766-25G
77333-676EA 239.99 USD
77333-676 77357-830 77369-682 77390-632
2,2-Difluoro-1-(3-methoxy-4-methylphenyl)ethanone ≥97%
2,2-Difluoro-1-(3-methoxy-4-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764194
CAS Number: 2229389-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR