1-(4-Butylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71714-500MG 71714-1G 71714-5G 71714-25G
77333-670EA 210.12 USD
77333-670 77357-824 77369-678 77390-626
1-(4-Butylphenyl)-2,2-difluoroethanone ≥97%
1-(4-Butylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O
MW: 212.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618096
CAS Number: 1352206-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR