1-(2,4-Dimethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71713-500MG 71713-1G 71713-5G 71713-25G
77333-668EA 210.12 USD
77333-668 77357-822 77369-676 77390-624
1-(2,4-Dimethylphenyl)-2,2-difluoroethanone ≥95%
1-(2,4-Dimethylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O
MW: 184.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920819
CAS Number: 1352214-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR