2,2-Difluoro-1-mesitylethanone ≥97%

Supplier: AOBChem USA

Warning

71701-500MG 71701-1G 71701-5G
77337-594EA 424.36 USD
77337-594 77357-812 77381-734
2,2-Difluoro-1-mesitylethanone ≥97%
2,2-Difluoro-1-mesitylethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
CAS Number: 1692847-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR