2-(2,2-Difluoroacetyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

71696-500MG 71696-1G 71696-25G 71696-5G
77333-658EA 305.91 USD
77333-658 77357-808 77390-614 77381-730
2-(2,2-Difluoroacetyl)benzonitrile ≥97%
2-(2,2-Difluoroacetyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₅F₂NO
Storage Temperature: Ambient
CAS Number: 1824268-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR