2,2-Difluoro-1-(3-hydroxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71687-500MG 71687-1G 71687-5G
77339-252EA 570.62 USD
77339-252 77357-806 77381-728
2,2-Difluoro-1-(3-hydroxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(3-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764184
CAS Number: 2228395-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR