1-(3-Bromo-5-methylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71682-500MG 71682-1G 71682-5G
77339-148EA 554.14 USD
77339-148 77357-804 77381-726
1-(3-Bromo-5-methylphenyl)-2,2-difluoroethanone ≥97%
1-(3-Bromo-5-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O
MW: 249.052 g/mol
Storage Temperature: Ambient
CAS Number: 2229369-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR