1-(2,4-Dichlorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71668-500MG 71668-1G 71668-5G
77336-948EA 400.67 USD
77336-948 77357-800 77381-722
1-(2,4-Dichlorophenyl)-2,2-difluoroethanone ≥97%
1-(2,4-Dichlorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄Cl₂F₂O
MW: 225.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28105065
CAS Number: 121412-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR