1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71650-500MG 71650-1G 71650-5G
77337-590EA 424.36 USD
77337-590 77357-796 77381-720
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-difluoroethanone ≥97%
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₈F₂O₃
MW: 214.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045087
CAS Number: 1567105-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR