2,2-Difluoro-1-(4-isobutylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

71647-500MG 71647-1G 71647-5G
77336-946EA 400.67 USD
77336-946 77357-794 77381-718
2,2-Difluoro-1-(4-isobutylphenyl)ethanone ≥97%
2,2-Difluoro-1-(4-isobutylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O
MW: 212.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103948
CAS Number: 1697618-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR