2,2-Difluoro-1-(4-pentylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71644-500MG 71644-1G 71644-5G
77337-588EA 424.36 USD
77337-588 77357-792 77381-716
2,2-Difluoro-1-(4-pentylphenyl)ethanone ≥95%
2,2-Difluoro-1-(4-pentylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆F₂O
MW: 226.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28104068
CAS Number: 1823859-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR