2,2-Difluoro-1-(1-methyl-1H-indol-5-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

71632-500MG 71632-1G 71632-5G
77339-146EA 554.14 USD
77339-146 77357-788 77381-712
2,2-Difluoro-1-(1-methyl-1H-indol-5-yl)ethanone ≥97%
2,2-Difluoro-1-(1-methyl-1H-indol-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₉F₂NO
MW: 209.192 g/mol
Storage Temperature: Ambient
CAS Number: 2228922-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR