4-(2,2-Difluoroacetyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

71601-500MG 71601-1G 71601-5G
77337-112EA 407.88 USD
77337-112 77357-780 77381-708
4-(2,2-Difluoroacetyl)benzonitrile ≥97%
4-(2,2-Difluoroacetyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD02169519
CAS Number: 1176519-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR