1-(2-Chlorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71597-500MG 71597-1G 71597-5G 71597-25G
77333-650EA 235.87 USD
77333-650 77357-778 77369-660 77390-606
1-(2-Chlorophenyl)-2,2-difluoroethanone ≥95%
1-(2-Chlorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O
MW: 190.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111889
CAS Number: 653-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR