2,2-Difluoro-1-(o-tolyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71592-500MG 71592-1G 71592-5G
77340-060EA 740.57 USD
77340-060 77357-772 77381-702
2,2-Difluoro-1-(o-tolyl)ethanone ≥95%
2,2-Difluoro-1-(o-tolyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618123
CAS Number: 703-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR