1-(Benzo[d][1,3]dioxol-5-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71591-500MG 71591-1G 71591-5G
77336-940EA 400.67 USD
77336-940 77357-770 77381-700
1-(Benzo[d][1,3]dioxol-5-yl)-2,2-difluoroethanone ≥97%
1-(Benzo[d][1,3]dioxol-5-yl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆F₂O₃
MW: 200.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13172860
CAS Number: 437552-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR