1-([1,1'-Biphenyl]-4-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71582-500MG 71582-1G 71582-5G 71582-25G
77333-642EA 210.12 USD
77333-642 77357-764 77369-654 77390-600
1-([1,1'-Biphenyl]-4-yl)-2,2-difluoroethanone ≥95%
1-([1,1'-Biphenyl]-4-yl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O
MW: 232.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618114
CAS Number: 75524-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR