2,2-Difluoro-1-(p-tolyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

71578-500MG 71578-1G 71578-5G 71578-25G
77333-638EA 184.37 USD
77333-638 77347-332 77369-650 77390-596
2,2-Difluoro-1-(p-tolyl)ethanone ≥97%
2,2-Difluoro-1-(p-tolyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618098
CAS Number: 704-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR