2,2-Difluoro-1-(4-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
71577-500MG
71577-1G
71577-5G
71577-25G
77333-636EA
210.12
USD
77333-636
77357-760
77369-648
77390-594
2,2-Difluoro-1-(4-methoxyphenyl)ethanone ≥97%
2,2-Difluoro-1-(4-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₈F₂O₂ MW: 186.155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13172721 CAS Number: 114829-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...