3-Chloro-4-iodo-5-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

71566-500MG 71566-1G 71566-5G 71566-25G
77333-634EA 247.2 USD
77333-634 77357-758 77369-646 77390-592
3-Chloro-4-iodo-5-methoxypyridine ≥97%
3-Chloro-4-iodo-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClINO
MW: 269.4674 g/mol
Storage Temperature: Ambient
CAS Number: 1261536-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR