1-Bromo-2-chloro-3,4-difluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

71549-500MG 71549-1G 71549-5G 71549-25G
77333-632EA 204.97 USD
77333-632 77357-756 77369-644 77390-590
1-Bromo-2-chloro-3,4-difluoro-5-iodobenzene ≥97%
1-Bromo-2-chloro-3,4-difluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆HBrClF₂I
MW: 353.3304 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764164
CAS Number: 2901107-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR