(2-(Benzyloxy)-5-ethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

71529-500MG 71529-1G 71529-5G 71529-25G
77333-628EA 143.17 USD
77333-628 77345-014 77369-642 77390-588
(2-(Benzyloxy)-5-ethylphenyl)(methyl)sulfane ≥97%
(2-(Benzyloxy)-5-ethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₆H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD34764161
CAS Number: 2807455-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR