2-Chloro-1-fluoro-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

71518-500MG 71518-1G 71518-5G 71518-25G
77333-626EA 187.46 USD
77333-626 77357-752 77369-640 77390-586
2-Chloro-1-fluoro-4-isopropoxybenzene ≥97%
2-Chloro-1-fluoro-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD29044095
CAS Number: 1369889-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR