2-(3-Chloro-2-fluoro-5-methoxyphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

71516-500MG 71516-1G 71516-5G 71516-25G
77333-624EA 276.04 USD
77333-624 77357-750 77369-638 77390-584
2-(3-Chloro-2-fluoro-5-methoxyphenyl)propan-2-ol ≥97%
2-(3-Chloro-2-fluoro-5-methoxyphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28531693
CAS Number: 1783704-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR