2-Chloro-1-fluoro-4-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

71498-500MG 71498-1G 71498-5G 71498-25G
77333-622EA 141.11 USD
77333-622 77344-920 77369-636 77390-582
2-Chloro-1-fluoro-4-propoxybenzene ≥95%
2-Chloro-1-fluoro-4-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD29044049
CAS Number: 1862372-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR