2-Bromo-1,3,5-trifluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

71495-500MG 71495-1G 71495-5G 71495-25G
77333-620EA 141.11 USD
77333-620 77344-918 77369-634 77390-580
2-Bromo-1,3,5-trifluoro-4-methoxybenzene ≥97%
2-Bromo-1,3,5-trifluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD28746652
CAS Number: 1805474-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR