5-Bromo-4-fluoro-2,3'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

71480-500MG 71480-1G 71480-5G 71480-25G
77333-616EA 161.71 USD
77333-616 77345-988 77369-628 77390-576
5-Bromo-4-fluoro-2,3'-dimethyl-1,1'-biphenyl ≥97%
5-Bromo-4-fluoro-2,3'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.1474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764144
CAS Number: 2901082-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR